Assay Details
Gene Symbol: PTGES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261168.1 | NP_001248097.1 | 237 | 86 | ||
GenBank mRNA | JU477928.1 | - | 237 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
