Assay Details
| Gene Symbol: CRYGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265732.2 | NP_001252661.1 | 292 | 70 | ||
| GenBank mRNA | JV635692.1 | - | 292 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRYGD| Gene Symbol: CRYGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265732.2 | NP_001252661.1 | 292 | 70 | ||
| GenBank mRNA | JV635692.1 | - | 292 | 70 | ||
Set Membership: |
|