Assay Details
Gene Symbol: CHST7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265790.1 | NP_001252719.1 | 1666 | 78 | ||
GenBank mRNA | JV600217.1 | - | 1666 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
