Assay Details
| Gene Symbol: JUN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265850.2 | NP_001252779.1 | 1824 | 81 | ||
| GenBank mRNA | JU334399.1 | - | 1852 | 81 | ||
| JV046989.1 | - | 1828 | 81 | |||
| JV580947.1 | - | 1831 | 81 | |||
More Information
Set Membership: |
|

JUN
Genomic Map