Assay Details
| Gene Symbol: HMOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266069.1 | NP_001252998.1 | 859 | 92 | ||
| XM_015125484.2 | XP_014980970.1 | 933 | 92 | |||
| XM_015125485.2 | XP_014980971.1 | 898 | 92 | |||
| XM_028840477.1 | XP_028696310.1 | 960 | 92 | |||
| GenBank mRNA | JU321669.1 | - | 882 | 92 | ||
| JU475384.1 | - | 825 | 92 | |||
| JV045002.1 | - | 859 | 92 | |||
More Information
Set Membership: |
|

HMOX2
Genomic Map