Assay Details
| Gene Symbol: RPL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266101.1 | NP_001253030.1 | 246 | 94 | ||
| XM_015146216.2 | XP_015001702.1 | 255 | 94 | |||
| XM_028852253.1 | XP_028708086.1 | 270 | 94 | |||
| GenBank mRNA | JU319968.1 | - | 246 | 94 | ||
| JV489538.1 | - | 223 | 94 | |||
More Information
Set Membership: |
|

RPL8
Genomic Map