Assay Details
| Gene Symbol: PMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266305.1 | NP_001253234.1 | 332 | 65 | ||
| GenBank mRNA | JU335038.1 | - | 332 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PMP2| Gene Symbol: PMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266305.1 | NP_001253234.1 | 332 | 65 | ||
| GenBank mRNA | JU335038.1 | - | 332 | 65 | ||
Set Membership: |
|