Assay Details
| Gene Symbol: SSNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266384.2 | NP_001253313.1 | 126 | 70 | ||
| GenBank mRNA | JU336879.1 | - | 69 | 70 | ||
| JV045613.1 | - | 126 | 70 | |||
| JV664288.1 | - | 135 | 70 | |||
More Information
Set Membership: |
|

SSNA1
Genomic Map