Assay Details
| Gene Symbol: PPP1R8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266462.1 | NP_001253391.1 | 62 | 80 | ||
| XM_028845627.1 | XP_028701460.1 | 88 | 80 | |||
| GenBank mRNA | JU319859.1 | - | 91 | 80 | ||
| JV045215.1 | - | 62 | 80 | |||
| JV654991.1 | - | 105 | 80 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028845627.1 transcripts.
Set Membership: |
|

PPP1R8
Genomic Map