Assay Details
| Gene Symbol: ACAA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266733.1 | NP_001253662.1 | 1263 | 110 | ||
| GenBank mRNA | JU322786.1 | - | 992 | 110 | ||
| JU473986.1 | - | 1262 | 110 | |||
| JV045913.1 | - | 1263 | 110 | |||
More Information
Set Membership: |
|

ACAA2
Genomic Map