Assay Details
| Gene Symbol: RGMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266917.1 | NP_001253846.1 | 757 | 94 | ||
| XM_028850456.1 | XP_028706289.1 | 2000 | 94 | |||
| XM_028850457.1 | XP_028706290.1 | 996 | 94 | |||
| XM_028850458.1 | XP_028706291.1 | 759 | 94 | |||
| XM_028850459.1 | XP_028706292.1 | 649 | 94 | |||
| GenBank mRNA | JU321454.1 | - | 757 | 94 | ||
| JU335545.1 | - | 844 | 94 | |||
| JV047710.1 | - | 935 | 94 | |||
More Information
Set Membership: |
|

RGMA
Genomic Map