Assay Details
Gene Symbol: LXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266988.1 | NP_001253917.1 | 309 | 83 | ||
GenBank mRNA | JU335536.1 | - | 312 | 83 | ||
JU476463.1 | - | 309 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
