Assay Details
| Gene Symbol: LSM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261364.1 | NP_001248293.1 | 180 | 105 | ||
| GenBank mRNA | JU334019.1 | - | 180 | 105 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LSM3| Gene Symbol: LSM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261364.1 | NP_001248293.1 | 180 | 105 | ||
| GenBank mRNA | JU334019.1 | - | 180 | 105 | ||
Set Membership: |
|