Assay Details
| Gene Symbol: AKR7A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265698.1 | NP_001252627.1 | 926 | 104 | ||
| GenBank mRNA | JU323316.1 | - | 934 | 104 | ||
| JU323317.1 | - | 934 | 104 | |||
| JU475012.1 | - | 930 | 104 | |||
| JU477603.1 | - | 930 | 104 | |||
| JV664234.1 | - | 945 | 104 | |||
| JV667234.1 | - | 926 | 104 | |||
More Information
Set Membership: |
|

AKR7A2
Genomic Map