Assay Details
Gene Symbol: HSPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266596.1 | NP_001253525.1 | 1265 | 86 | ||
GenBank mRNA | JU320096.1 | - | 1186 | 86 | ||
JU471192.1 | - | 1265 | 86 | |||
JV638117.1 | - | 1272 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
