Assay Details
| Gene Symbol: DPH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260520.1 | NP_001247449.1 | 194 | 108 | ||
| GenBank mRNA | JV046815.1 | - | 194 | 108 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DPH3| Gene Symbol: DPH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260520.1 | NP_001247449.1 | 194 | 108 | ||
| GenBank mRNA | JV046815.1 | - | 194 | 108 | ||
Set Membership: |
|