Assay Details
Gene Symbol: NSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261761.1 | NP_001248690.1 | 916 | 150 | ||
GenBank mRNA | JU320656.1 | - | 968 | 150 | ||
JU473972.1 | - | 881 | 150 | |||
JV044945.1 | - | 916 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
