Assay Details
| Gene Symbol: TXNDC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266161.1 | NP_001253090.1 | 1115 | 150 | ||
| GenBank mRNA | JU321439.1 | - | 1154 | 151 | ||
| JU475809.1 | - | 1071 | 151 | |||
| JV045689.1 | - | 1115 | 150 | |||
| JV567081.1 | - | 1227 | 151 | |||
More Information
Set Membership: |
|

TXNDC12
Genomic Map