Assay Details
Gene Symbol: SMIM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260616.2 | NP_001247545.1 | 231 | 121 | ||
XM_015134843.2 | XP_014990329.1 | 395 | 121 | |||
XM_015134848.2 | XP_014990334.1 | 359 | 121 | |||
XM_015134855.2 | XP_014990341.1 | 1162 | 121 | |||
XM_028844544.1 | XP_028700377.1 | 224 | 121 | |||
GenBank mRNA | JU320349.1 | - | 218 | 121 | ||
JU474205.1 | - | 202 | 121 | |||
JU474206.1 | - | 337 | 121 | |||
JV046099.1 | - | 118 | 121 | |||
JV490358.1 | - | 156 | 121 | |||
JV529385.1 | - | 231 | 121 | |||
JV704415.1 | - | 205 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
