Assay Details
Gene Symbol: RBM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002801750.3 | XP_002801796.1 | 418 | 73 | ||
XM_015150144.2 | XP_015005630.1 | 444 | 73 | |||
GenBank mRNA | JU337394.1 | - | 371 | 73 | ||
JU478091.1 | - | 375 | 73 | |||
JU478092.1 | - | 375 | 73 | |||
JU478093.1 | - | 375 | 73 | |||
JU478094.1 | - | 375 | 73 | |||
JV044011.1 | - | 371 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
