Assay Details
Gene Symbol: DUSP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261330.1 | NP_001248259.1 | 406 | 74 | ||
GenBank mRNA | JU321544.1 | - | 376 | 74 | ||
JU474430.1 | - | 339 | 74 | |||
JU474431.1 | - | 339 | 74 | |||
JU474432.1 | - | 339 | 74 | |||
JU474433.1 | - | 339 | 74 | |||
JU474434.1 | - | 339 | 74 | |||
JU474435.1 | - | 339 | 74 | |||
JV043868.1 | - | 304 | 74 | |||
JV043869.1 | - | 406 | 74 | |||
JV572356.1 | - | 407 | 74 | |||
JV639301.1 | - | 431 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001261330.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
