Assay Details
Gene Symbol: GET4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133149.2 | XP_014988635.2 | 418 | 61 | ||
XM_015133150.2 | XP_014988636.2 | 416 | 61 | |||
XM_015133151.2 | XP_014988637.1 | 897 | 61 | |||
XM_028845474.1 | XP_028701307.1 | 1186 | 61 | |||
XM_028845475.1 | XP_028701308.1 | 1575 | 61 | |||
GenBank mRNA | JU334940.1 | - | 396 | 61 | ||
JU475942.1 | - | 396 | 61 | |||
JV047334.1 | - | 330 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
