Assay Details
| Gene Symbol: OBP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834654.1 | XP_028690487.1 | 205 | 89 | ||
| XM_028834655.1 | XP_028690488.1 | 205 | 89 | |||
| GenBank mRNA | JV542832.1 | - | 75 | 89 | ||
| JV614737.1 | - | 75 | 89 | |||
More Information
Set Membership: |
|

OBP2B
Genomic Map