Assay Details
Gene Symbol: SH3BGRL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015136833.2 | XP_014992319.1 | 658 | 107 | ||
XM_028847398.1 | XP_028703231.1 | 379 | 107 | |||
GenBank mRNA | JU336627.1 | - | 476 | 107 | ||
JV043491.1 | - | 424 | 107 | |||
JV602370.1 | - | 487 | 107 | |||
JV724683.1 | - | 486 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
