Assay Details
| Gene Symbol: SLC25A29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002805208.3 | XP_002805254.3 | 465 | 60 | ||
| XM_015144483.2 | XP_014999969.2 | 1857 | 60 | |||
| XM_028851826.1 | XP_028707659.1 | 1296 | 60 | |||
| XM_028851827.1 | XP_028707660.1 | 1200 | 60 | |||
| XM_028851828.1 | XP_028707661.1 | 1076 | 60 | |||
| XM_028851829.1 | XP_028707662.1 | 1198 | 60 | |||
| XM_028851830.1 | XP_028707663.1 | 1159 | 60 | |||
| XM_028851831.1 | XP_028707664.1 | 334 | 60 | |||
| GenBank mRNA | JU332878.1 | - | 329 | 60 | ||
| JU480006.1 | - | 83 | 60 | |||
| JV527783.1 | - | 290 | 60 | |||
More Information
Set Membership: |
|

SLC25A29
Genomic Map