Assay Details
| Gene Symbol: LZTS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149421.2 | XP_015004907.1 | 797 | 94 | ||
| XM_015149423.2 | XP_015004909.1 | 841 | 94 | |||
| XM_015149424.2 | XP_015004910.1 | 743 | 94 | |||
| XM_028828264.1 | XP_028684097.1 | 2364 | 94 | |||
| GenBank mRNA | JU319700.1 | - | 745 | 94 | ||
| JV565510.1 | - | 1436 | 94 | |||
| JV600381.1 | - | 1455 | 94 | |||
More Information
Set Membership: |
|

LZTS3
Genomic Map