Assay Details
| Gene Symbol: ZNF563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028839170.1 | XP_028695003.1 | 1260 | 101 | ||
| XM_028839171.1 | XP_028695004.1 | 1262 | 101 | |||
| GenBank mRNA | JU480903.1 | - | 91 | 101 | ||
More Information
Set Membership: |
|

ZNF563
Genomic Map