Assay Details
| Gene Symbol: ZNF563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028839170.1 | XP_028695003.1 | 1322 | 102 | ||
| GenBank mRNA | JU480903.1 | - | 153 | 102 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF563| Gene Symbol: ZNF563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028839170.1 | XP_028695003.1 | 1322 | 102 | ||
| GenBank mRNA | JU480903.1 | - | 153 | 102 | ||
Set Membership: |
|