Assay Details
| Gene Symbol: SGPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143868.2 | XP_014999354.1 | 912 | 116 | ||
| GenBank mRNA | JU327053.1 | - | 513 | 116 | ||
| JU494248.1 | - | 497 | 116 | |||
More Information
Set Membership: |
|

SGPP1
Genomic Map