Assay Details
| Gene Symbol: OGFOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028841321.1 | XP_028697154.1 | 409 | 93 | ||
| XM_028841322.1 | XP_028697155.1 | 609 | 93 | |||
| GenBank mRNA | JU335281.1 | - | 384 | 93 | ||
| JV718094.1 | - | 372 | 93 | |||
More Information
Set Membership: |
|

OGFOD1
Genomic Map