Assay Details
| Gene Symbol: GPRC5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028841432.1 | XP_028697265.1 | 1223 | 59 | ||
| XM_028841433.1 | XP_028697266.1 | 1564 | 59 | |||
| XM_028841434.1 | XP_028697267.1 | 1104 | 59 | |||
| GenBank mRNA | JV044395.1 | - | 1140 | 59 | ||
| JV653563.1 | - | 1053 | 59 | |||
More Information
Set Membership: |
|

GPRC5B
Genomic Map