Assay Details
| Gene Symbol: FKBP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001104575.4 | XP_001104575.2 | 1167 | 93 | ||
| XM_002798556.3 | XP_002798602.1 | 327 | 93 | |||
| GenBank mRNA | JV062821.1 | - | 216 | 93 | ||
| JV716456.1 | - | 201 | 93 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002798556.3 transcripts.
Set Membership: |
|

FKBP11
Genomic Map