Assay Details
Gene Symbol: LOC114669852 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842799.1 | XP_028698632.1 | 557 | 60 | ||
GenBank mRNA | JU476522.1 | - | 523 | 60 | ||
JV066501.1 | - | 536 | 60 |
Gene Symbol: RCN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015124582.2 | XP_014980068.2 | 724 | 60 | ||
GenBank mRNA | JV551913.1 | - | 186 | 60 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
