Assay Details
Gene Symbol: NDUFAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141982.2 | XP_014997468.1 | 1719 | 69 | ||
XM_015141983.2 | XP_014997469.1 | 1216 | 69 | |||
GenBank mRNA | JU323584.1 | - | 1211 | 69 | ||
JU471855.1 | - | 1215 | 69 | |||
JV047247.1 | - | 1222 | 69 | |||
JV652989.1 | - | 601 | 69 | |||
JV667313.1 | - | 1145 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
