Assay Details
| Gene Symbol: FXYD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146885.2 | XP_015002371.1 | 304 | 85 | ||
| XM_015146886.2 | XP_015002372.1 | 277 | 85 | |||
| XM_015146887.2 | XP_015002373.1 | 298 | 85 | |||
| XM_015146888.2 | XP_015002374.1 | 271 | 85 | |||
More Information
Set Membership: |
|

FXYD4
Genomic Map