Assay Details
Gene Symbol: RNF135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015118995.2 | XP_014974481.2 | 2001 | 74 | ||
XM_015118996.2 | XP_014974482.2 | 1874 | 74 | |||
XM_015118997.2 | XP_014974483.1 | 1940 | 74 | |||
XM_028836197.1 | XP_028692030.1 | 2388 | 74 | |||
GenBank mRNA | JU325286.1 | - | 284 | 74 | ||
JU494785.1 | - | 637 | 74 | |||
JV725018.1 | - | 403 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
