Assay Details
Gene Symbol: LUZP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015128001.2 | XP_014983487.1 | 390 | 67 | ||
GenBank mRNA | JV062724.1 | - | 286 | 67 | ||
JV653110.1 | - | 230 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
