Assay Details
| Gene Symbol: PRKACA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123119.2 | XP_014978605.1 | 748 | 55 | ||
| XM_015123120.2 | XP_014978606.1 | 557 | 55 | |||
| GenBank mRNA | JU320957.1 | - | 583 | 55 | ||
| JU473759.1 | - | 571 | 55 | |||
| JV044411.1 | - | 576 | 55 | |||
More Information
Set Membership: |
|

PRKACA
Genomic Map