Assay Details
Gene Symbol: PAFAH1B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257739.1 | NP_001244668.1 | 552 | 119 | ||
XM_015115708.2 | XP_014971194.1 | 513 | 119 | |||
XM_028832602.1 | XP_028688435.1 | 601 | 119 | |||
XM_028832603.1 | XP_028688436.1 | 490 | 119 | |||
GenBank mRNA | JU334729.1 | - | 552 | 119 | ||
JU475776.1 | - | 540 | 119 | |||
JU475777.1 | - | 540 | 119 | |||
JV045384.1 | - | 502 | 119 | |||
JV045385.1 | - | 502 | 119 | |||
JV618100.1 | - | 553 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
