Assay Details
| Gene Symbol: LOC719811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126871.2 | XP_014982357.1 | 905 | 60 | ||
| GenBank mRNA | JU319663.1 | - | 812 | 60 | ||
| JV046852.1 | - | 771 | 60 | |||
| JV649404.1 | - | 878 | 60 | |||
| JV681752.1 | - | 877 | 60 | |||
More Information
Set Membership: |
|

LOC719811
Genomic Map