Assay Details
| Gene Symbol: SRY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032836.1 | NP_001028008.1 | 520 | 75 | ||
| GenBank mRNA | AF322901.1 | - | 520 | 75 | ||
| FJ527022.1 | - | 542 | 75 | |||
| JV069553.1 | - | 350 | 75 | |||
More Information
Set Membership: |
|

SRY
Genomic Map