Assay Details
| Gene Symbol: RPRML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194690.4 | NP_001181619.1 | 327 | 98 | ||
| GenBank mRNA | JU334450.1 | - | 197 | 98 | ||
| JV667258.1 | - | 331 | 98 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPRML| Gene Symbol: RPRML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194690.4 | NP_001181619.1 | 327 | 98 | ||
| GenBank mRNA | JU334450.1 | - | 197 | 98 | ||
| JV667258.1 | - | 331 | 98 | |||