Assay Details
| Gene Symbol: IGBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195789.1 | NP_001182718.1 | 755 | 72 | ||
| XM_015127616.2 | XP_014983102.1 | 1069 | 72 | |||
| GenBank mRNA | CN803525.1 | - | 737 | 72 | ||
| CO581702.1 | - | 531 | 72 | |||
| JU322837.1 | - | 894 | 72 | |||
| JU472917.1 | - | 923 | 72 | |||
| JV044141.1 | - | 786 | 72 | |||
| JV044142.1 | - | 786 | 72 | |||
More Information
Set Membership: |
|

IGBP1
Genomic Map