Assay Details
| Gene Symbol: MRPL36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258097.1 | NP_001245026.1 | 36 | 80 | ||
| GenBank mRNA | JU335975.1 | - | 57 | 80 | ||
| JU476856.1 | - | 36 | 80 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001258097.1 transcripts.
Set Membership: |
|

MRPL36
Genomic Map