Assay Details
Gene Symbol: ATP5PB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260679.2 | NP_001247608.1 | 228 | 71 | ||
GenBank mRNA | JU334908.1 | - | 198 | 71 | ||
JU475913.1 | - | 93 | 71 | |||
JV044839.1 | - | 229 | 71 | |||
JV470977.1 | - | 193 | 71 | |||
JV667200.1 | - | 159 | 71 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
