Assay Details
| Gene Symbol: RPS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266796.1 | NP_001253725.1 | 369 | 90 | ||
| XM_015111622.1 | XP_014967108.1 | 514 | 90 | |||
| GenBank mRNA | JV068073.1 | - | 390 | 90 | ||
| JV516213.1 | - | 451 | 90 | |||
| JV635878.1 | - | 369 | 90 | |||
More Information
Set Membership: |
|

RPS7
Genomic Map