Assay Details
Gene Symbol: DEFB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128851.2 | NP_001122323.2 | 137 | 88 | ||
GenBank mRNA | EU090141.1 | - | 123 | 88 | ||
EU090142.1 | - | 123 | 88 | |||
EU090143.1 | - | 123 | 88 | |||
JU481566.1 | - | 88 |
Gene Symbol: LOC114675439 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842691.1 | XP_028698524.1 | 188 | 88 |
Gene Symbol: LOC114680396 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028853174.1 | XP_028709007.1 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
