Assay Details
| Gene Symbol: ATP6V1G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193602.1 | NP_001180531.1 | 509 | 104 | ||
| GenBank mRNA | CB311811.1 | - | 512 | 104 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP6V1G1| Gene Symbol: ATP6V1G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193602.1 | NP_001180531.1 | 509 | 104 | ||
| GenBank mRNA | CB311811.1 | - | 512 | 104 | ||
Set Membership: |
|