Assay Details
Gene Symbol: BOLA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194470.3 | NP_001181399.2 | 519 | 130 | ||
XM_015125897.2 | XP_014981383.2 | 595 | 130 | |||
XM_028840539.1 | XP_028696372.1 | 153 | 130 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194470.3, XM_015125897.2, XM_028840539.1 transcripts.
Set Membership: |
![]() ![]() |
