Assay Details
Gene Symbol: DEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257528.1 | NP_001244457.1 | 1512 | 122 | ||
XM_028846797.1 | XP_028702630.1 | 1622 | 122 | |||
GenBank mRNA | JU321533.1 | - | 1523 | 122 | ||
JU321535.1 | - | 1512 | 122 | |||
JV044596.1 | - | 1529 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
